Thermo Scientific Chemicals

Tris(2-carboxyethyl)phosphine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 363830010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Tris(2-carboxyethyl)phosphine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 363830010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 363830010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51805-45-9
IUPAC Name
3-[bis(2-carboxylatoethyl)phosphanyl]propanoate
Molecular Formula
C9H12O6P
InChI Key
PZBFGYYEXUXCOF-UHFFFAOYSA-K
SMILES
[O-]C(=O)CCP(CCC([O-])=O)CCC([O-])=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder or crystals
Titration with NaOH
>=97.5 %
Solubility
(50mg/ml water) Clear colorless
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text