Thermo Scientific Chemicals

Tris(hydroxymethyl)phosphine, 95%, Thermo Scientific Chemicals

Catalog number: 363840010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Tris(hydroxymethyl)phosphine, 95%, Thermo Scientific Chemicals

Catalog number: 363840010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 363840010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2767-80-8
IUPAC Name
[bis(hydroxymethyl)phosphanyl]methanol
Molecular Formula
C3H9O3P
InChI Key
JMXMXKRNIYCNRV-UHFFFAOYSA-N
SMILES
OCP(CO)CO
Appearance (Form)
Powder or crystals and/or chunks
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
NMR
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text