Thermo Scientific Chemicals

1,2-Bis(dicyclohexylphosphino)ethane, 98%, Thermo Scientific Chemicals

Catalog number: 363850050
5 g, Each
Thermo Scientific Chemicals

1,2-Bis(dicyclohexylphosphino)ethane, 98%, Thermo Scientific Chemicals

Catalog number: 363850050
5 g, Each
Quantity
Catalog number: 363850050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
23743-26-2
IUPAC Name
dicyclohexyl[2-(dicyclohexylphosphanyl)ethyl]phosphane
Molecular Formula
C26H48P2
InChI Key
BOUYBUIVMHNXQB-UHFFFAOYSA-N
SMILES
C(CP(C1CCCCC1)C1CCCCC1)P(C1CCCCC1)C1CCCCC1
Appearance (Color)
White to almost white
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
NMR
>=97.5 %
Melting point
92°C to 98°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text