Thermo Scientific Chemicals

chloro(1,5-cyclooctadiene)iridium(I) dimer, Thermo Scientific Chemicals

Catalog number: 363960010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

chloro(1,5-cyclooctadiene)iridium(I) dimer, Thermo Scientific Chemicals

Catalog number: 363960010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 363960010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12112-67-3
IUPAC Name
diiridium(1+) bis((1Z,5Z)-cycloocta-1,5-diene) dichloride
Molecular Formula
C16H24Cl2Ir2
InChI Key
ZFOUDQNHNLDNLD-MIXQCLKLSA-L
SMILES
[Cl-].[Cl-].[Ir+].[Ir+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
Appearance (Form)
Powder
Infrared spectrum
Conforms
Iridium (Ir)
>=56.4 %
Appearance (Color)
Orange to red

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text