chloro(1,5-cyclooctadiene)iridium(I) dimer, Thermo Scientific Chemicals
chloro(1,5-cyclooctadiene)iridium(I) dimer, Thermo Scientific Chemicals
Thermo Scientific Chemicals

chloro(1,5-cyclooctadiene)iridium(I) dimer, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 mg
1 g
Packaging:
Glass bottle
Catalog number 363960050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namediiridium(1+) bis((1Z,5Z)-cycloocta-1,5-diene) dichloride
Molecular FormulaC16H24Cl2Ir2
InChI KeyZFOUDQNHNLDNLD-MIXQCLKLSA-L
SMILES[Cl-].[Cl-].[Ir+].[Ir+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
Molecular Weight (g/mol)671.70
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Iridium (Ir)>=56.4 %
Appearance (Color)Orange to red
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only