Thermo Scientific Chemicals

Methyl 6-bromo-2-naphthoate, 98%, Thermo Scientific Chemicals

Catalog number: 364140050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl 6-bromo-2-naphthoate, 98%, Thermo Scientific Chemicals

Catalog number: 364140050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 364140050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
33626-98-1
IUPAC Name
methyl 6-bromonaphthalene-2-carboxylate
Molecular Formula
C12H9BrO2
InChI Key
JEUBRLPXJZOGPX-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C2C=C(Br)C=CC2=C1
Appearance (Color)
Off-white to cream to light brown
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
GC
>=97.5 %
Melting point
118°C to 126°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text