Dichloromethane, 99.8%, for HPLC, stabilized with methanol
Dichloromethane, 99.8%, for HPLC, stabilized with methanol
Dichloromethane, 99.8%, for HPLC, stabilized with methanol
Dichloromethane, 99.8%, for HPLC, stabilized with methanol
Thermo Scientific Chemicals

Dichloromethane, 99.8%, for HPLC, stabilized with methanol

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 364230010
Price (USD)
65.65
Special offer
Online exclusive
77.20
Save 11.55 (15%)
Each
-
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
65.65
Special offer
Online exclusive
77.20
Save 11.55 (15%)
Each
Add to cart
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=99.8 %
UVat 233 nm A: =<1
UV1 cm cell vs HPLC-grade water
Free chlorine=<0.0005 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for HPLC
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473084Certificate of AnalysisJun 11, 2025364230025, 364230010
1991832Certificate of AnalysisJan 27, 2025364230025, 364230010
1991830Certificate of AnalysisJan 27, 2025364230025, 364230010
1993027Certificate of AnalysisJan 27, 2025364230025, 364230010
1905360Certificate of AnalysisJan 27, 2025364230025, 364230010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection