Dichloromethane, 99.8%, for HPLC, stabilized with methanol, Thermo Scientific Chemicals
Dichloromethane, 99.8%, for HPLC, stabilized with methanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dichloromethane, 99.8%, for HPLC, stabilized with methanol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number 364230025
Price (USD)/ Each
126.65
Online exclusive
140.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
126.65
Online exclusive
140.00 
Save 13.35 (10%)
Add to cart
Dichloromethane, 99.8%, for HPLC, stabilized with methanol, Thermo Scientific Chemicals
Catalog number364230025
Price (USD)/ Each
126.65
Online exclusive
140.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=99.8 %
UVat 233 nm A: =<1
UV1 cm cell vs HPLC-grade water
Free chlorine=<0.0005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for HPLC
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only