1,4-Dioxane, 99.5%, Extra Dry over Molecular Sieve, stabilized, AcroSeal™, Thermo Scientific Chemicals
1,4-Dioxane, 99.5%, Extra Dry over Molecular Sieve, stabilized, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Dioxane, 99.5%, Extra Dry over Molecular Sieve, stabilized, AcroSeal™, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
500 mL
Catalog number 364345000
Price (USD)/ Each
80.65
Online exclusive
89.20 
Save 8.55 (10%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
80.65
Online exclusive
89.20 
Save 8.55 (10%)
Add to cart
1,4-Dioxane, 99.5%, Extra Dry over Molecular Sieve, stabilized, AcroSeal™, Thermo Scientific Chemicals
Catalog number364345000
Price (USD)/ Each
80.65
Online exclusive
89.20 
Save 8.55 (10%)
-
Add to cart
Chemical Identifiers
CAS123-91-1
IUPAC Name1,4-dioxane
Molecular FormulaC4H8O2
InChI KeyRYHBNJHYFVUHQT-UHFFFAOYSA-N
SMILESC1COCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Acidity (CH3COOH)=<0.002 %
Stabilizer2 to 5 ppm BHT
Appearance (Color)Clear colorless
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only