1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 364380025
Price (USD)/ Each
297.65
Online exclusive
331.00 
Save 33.35 (10%)
-
Add to cart
Quantity:
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
297.65
Online exclusive
331.00 
Save 33.35 (10%)
Add to cart
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Catalog number364380025
Price (USD)/ Each
297.65
Online exclusive
331.00 
Save 33.35 (10%)
-
Add to cart
Chemical Identifiers
CAS872-50-4
IUPAC Name1-methylpyrrolidin-2-one
Molecular FormulaC5H9NO
InChI KeySECXISVLQFMRJM-UHFFFAOYSA-N
SMILESCN1CCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.5 %
Appearance (Form)Clear liquid
Infrared spectrumConforms
Water=<0.005 % (Coulometric)
Color scale=<50 APHA

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only