Thermo Scientific Chemicals

2,2,4-Trimethylpentane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 364671000
100 mL, Each
Thermo Scientific Chemicals

2,2,4-Trimethylpentane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 364671000
100 mL, Each
Quantity
Catalog number: 364671000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
540-84-1
IUPAC Name
2,2,4-trimethylpentane
Molecular Formula
C8H18
InChI Key
NHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILES
CC(C)CC(C)(C)C
Water
=<0.005 % (Coulometric)
Appearance (Form)
Liquid
GC
>=99.4 %
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text