Sodium pivalate hydrate, 99%, Thermo Scientific Chemicals
Sodium pivalate hydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium pivalate hydrate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 364710100
Price (USD)/ Each
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS143174-36-1
IUPAC Namesodium 2,2-dimethylpropanoate
Molecular FormulaC5H9NaO2
InChI KeySJRDNQOIQZOVQD-UHFFFAOYSA-M
SMILES[Na+].CC(C)(C)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Infrared spectrumConforms
Water=<7 % (K.F.)
Titration with HClO4>=97.5 % (On dry substance)
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only