Thermo Scientific Chemicals

2-Methylresorcinol, 98%, Thermo Scientific Chemicals

Catalog number: 364741000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Methylresorcinol, 98%, Thermo Scientific Chemicals

Catalog number: 364741000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 364741000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
608-25-3
IUPAC Name
2-methylbenzene-1,3-diol
Molecular Formula
C7H8O2
InChI Key
ZTMADXFOCUXMJE-UHFFFAOYSA-N
SMILES
CC1=C(O)C=CC=C1O
Appearance (Form)
Crystalline powder
GC
>=97.5 %
Melting point
118°C to 124°C
Appearance (Color)
Beige to pink to brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text