Thermo Scientific Chemicals

Guanidine hydrochloride, 99.5%, without anticaking agent, Thermo Scientific Chemicals

Catalog number: 364790025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Guanidine hydrochloride, 99.5%, without anticaking agent, Thermo Scientific Chemicals

Catalog number: 364790025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 364790025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50-01-1
IUPAC Name
hydrogen guanidine chloride
Molecular Formula
CH6ClN3
InChI Key
PJJJBBJSCAKJQF-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NC(N)=N
Infrared spectrum
Conforms to structure and standard
Identification A
Passes test
UV
at 260 nm A: =<0.030
Appearance (Color)
Colorless to white
Titration with HClO4
>=99.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text