Guanidine hydrochloride, 99.5%, without anticaking agent, Thermo Scientific Chemicals
Guanidine hydrochloride, 99.5%, without anticaking agent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Guanidine hydrochloride, 99.5%, without anticaking agent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 364795000
Price (USD)/ Each
197.65
Online exclusive
219.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
197.65
Online exclusive
219.00 
Save 21.35 (10%)
Add to cart
Guanidine hydrochloride, 99.5%, without anticaking agent, Thermo Scientific Chemicals
Catalog number364795000
Price (USD)/ Each
197.65
Online exclusive
219.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS50-01-1
IUPAC Namehydrogen guanidine chloride
Molecular FormulaCH6ClN3
InChI KeyPJJJBBJSCAKJQF-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC(N)=N
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms to structure and standard
Identification APasses test
UVat 260 nm A: =<0.030
Appearance (Color)Colorless to white
Titration with HClO4>=99.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only