Thermo Scientific Chemicals

Aluminum chloride, 99.999%, (trace metal basis), for analysis, anhydrous, powder, Thermo Scientific Chemicals

Catalog number: 364810200
20 g, Glass Bottle, Each
Thermo Scientific Chemicals

Aluminum chloride, 99.999%, (trace metal basis), for analysis, anhydrous, powder, Thermo Scientific Chemicals

Catalog number: 364810200
20 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 364810200
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7446-70-0
IUPAC Name
aluminium(3+) trichloride
Molecular Formula
AlCl3
InChI Key
VSCWAEJMTAWNJL-UHFFFAOYSA-K
SMILES
[Al+3].[Cl-].[Cl-].[Cl-]
Appearance (Form)
Powder
Iron (Fe)
=<5 ppm
Total trace metal impurities
=<0.001 %
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis, Lewis acid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text