Thermo Scientific Chemicals

Benzyltrimethylammonium bromide, 98+%, Thermo Scientific Chemicals

Catalog number: 364860250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Benzyltrimethylammonium bromide, 98+%, Thermo Scientific Chemicals

Catalog number: 364860250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 364860250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5350-41-4
IUPAC Name
benzyltrimethylazanium bromide
Molecular Formula
C10H16BrN
InChI Key
UUZYBYIOAZTMGC-UHFFFAOYSA-M
SMILES
[Br-].C[N+](C)(C)CC1=CC=CC=C1
Appearance (Form)
Crystalline powder
Appearance (Color)
White to cream
Infrared spectrum
Conforms
Melting point
224°C to 234°C
Titration Argentometric
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text