Thermo Scientific Chemicals

6-Hydroxyquinoline, 98%, Thermo Scientific Chemicals

Catalog number: 364931000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

6-Hydroxyquinoline, 98%, Thermo Scientific Chemicals

Catalog number: 364931000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 364931000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
580-16-5
IUPAC Name
quinolin-6-ol
Molecular Formula
C9H7NO
InChI Key
OVYWMEWYEJLIER-UHFFFAOYSA-N
SMILES
OC1=CC=C2N=CC=CC2=C1
Appearance (Color)
Beige to grey to brown
HPLC
>=97.5 %
Melting point
190°C to 195°C
Infrared spectrum
Conforms
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text