n-Heptane, for HPLC, (approx. 99% n-Heptane), Thermo Scientific Chemicals
n-Heptane, for HPLC, (approx. 99% n-Heptane), Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Heptane, for HPLC, (approx. 99% n-Heptane), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 364970025
Price (USD)/ Each
-
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
CAS142-82-5
IUPAC Nameheptane
Molecular FormulaC7H16
InChI KeyIMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILESCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after evaporation=<5 ppm
UVat 250 nm A: =<0.005
UVat 220 nm A: =<0.05
UVat 230 nm A: =<0.02
UVat 240 nm A: =<0.01
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only