Thermo Scientific Chemicals

Ruthenium(III) nitrosyl nitrate, in diluted aq. nitric acid solution, Thermo Scientific Chemicals

Catalog number: 365040250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

Ruthenium(III) nitrosyl nitrate, in diluted aq. nitric acid solution, Thermo Scientific Chemicals

Catalog number: 365040250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 365040250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
34513-98-9
IUPAC Name
λ²-ruthenium(2+) iminooxidanium trinitrate
Molecular Formula
N4O10Ru
InChI Key
WOSOOWIGVAKGOC-UHFFFAOYSA-N
SMILES
[Ru++].N#[O+].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Appearance (Color)
Clear orange to dark brown
Appearance (Form)
Solution
Assay
1.5 % Ru (typically)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text