2-Bromo-4'-cyanoacetophenone, 97%, Thermo Scientific Chemicals
2-Bromo-4'-cyanoacetophenone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-4'-cyanoacetophenone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 365360250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name4-(2-bromoacetyl)benzonitrile
Molecular FormulaC9H6BrNO
InChI KeyLJANCPRIUMHGJE-UHFFFAOYSA-N
SMILESBrCC(=O)C1=CC=C(C=C1)C#N
Molecular Weight (g/mol)224.06
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=96.0 %
Melting point90°C to 96°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only