Thermo Scientific Chemicals

Lithium metaborate, 99%, for analysis, Thermo Scientific Chemicals

Catalog number: 365711000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Lithium metaborate, 99%, for analysis, Thermo Scientific Chemicals

Catalog number: 365711000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 365711000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13453-69-5
IUPAC Name
tetraboron heptahydrate dilithium
Molecular Formula
B4H14Li2O7
InChI Key
DPYKRXYVXDYLEY-UHFFFAOYSA-N
SMILES
[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O
Calcium (Ca)
=<200 ppm
Iron (Fe)
=<50 ppm
Appearance (Color)
White
Appearance (Form)
Powder
Titration with HCl
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text