Thermo Scientific Chemicals

Aluminum oxide, for chromatography, acidic, Brockmann I, 40-300 μm, 60A, Thermo Scientific Chemicals

Catalog number: 365750025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Aluminum oxide, for chromatography, acidic, Brockmann I, 40-300 μm, 60A, Thermo Scientific Chemicals

Catalog number: 365750025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 365750025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1344-28-1
IUPAC Name
dialuminium(3+) trioxidandiide
Molecular Formula
Al2O3
InChI Key
PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Al+3].[Al+3]
Appearance (Form)
Powder
Sodium oxide (Na2O)
=<0.4 %
Specific surface area
185 to 225 m²/g
Appearance (Color)
White to off-white
Silicon dioxide (SiO2)
=<0.03 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for column chromatography
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text