Thermo Scientific Chemicals

2,2,2-Trifluoroethylhydrazine, 70 wt% aqueous solution, Thermo Scientific Chemicals

Catalog number: 365760050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2,2,2-Trifluoroethylhydrazine, 70 wt% aqueous solution, Thermo Scientific Chemicals

Catalog number: 365760050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 365760050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5042-30-8
IUPAC Name
(2,2,2-trifluoroethyl)hydrazine
Molecular Formula
C2H5F3N2
InChI Key
OPMFFAOEPFATTG-UHFFFAOYSA-N
SMILES
NNCC(F)(F)F
Appearance (Form)
Solution
Refractive index
1.3540 to 1.3640 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow
Titration with HClO4
60 to 70 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text