2,2,2-Trifluoroethylhydrazine, 70 wt% aqueous solution, Thermo Scientific Chemicals
2,2,2-Trifluoroethylhydrazine, 70 wt% aqueous solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2,2-Trifluoroethylhydrazine, 70 wt% aqueous solution, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 365760250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5042-30-8
IUPAC Name(2,2,2-trifluoroethyl)hydrazine
Molecular FormulaC2H5F3N2
InChI KeyOPMFFAOEPFATTG-UHFFFAOYSA-N
SMILESNNCC(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Refractive index1.3540 to 1.3640 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Titration with HClO460 to 70 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only