Thermo Scientific Chemicals

Dodecane, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 365770010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

Dodecane, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 365770010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 365770010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13475-82-6
IUPAC Name
2,2,4,6,6-pentamethylheptane
Molecular Formula
C12H26
InChI Key
VKPSKYDESGTTFR-UHFFFAOYSA-N
SMILES
CC(CC(C)(C)C)CC(C)(C)C
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless
Refractive index
1.4190 to 1.4230 (20°C, 589 nm)
Distillation range
170 to 195 °C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text