1x1 image pixel for data collection
Dodecane, mixture of isomers
Dodecane, mixture of isomers
Dodecane, mixture of isomers
Dodecane, mixture of isomers
Thermo Scientific Chemicals

Dodecane, mixture of isomers

Dodecane, C12H26, CAS Number-13475-82-6, heptane,2,2,4,6,6-pentamethyl, 2,2,4,6,6-pentamethyl-heptane, heptane, 2,2,4,6,6-pentamethyl, acmc-1c04u, vkpskydesgttfr-uhfffaoysa, permethyl 99a, 1L, 170.0 deg.C to 195.0 deg.C, CHEBI:77509, Undesignated, 0.7510g/mL, 236-757-, 44 deg.C, 170.34 | CAS: 13475-82-6 | C12H26 | 170.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 L
Packaging:
View all
Plastic drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 365770050
Price (USD)
-
Quantity:
5 L
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS13475-82-6
IUPAC Name2,2,4,6,6-pentamethylheptane
Molecular FormulaC12H26
InChI KeyVKPSKYDESGTTFR-UHFFFAOYSA-N
SMILESCC(CC(C)(C)C)CC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive index1.4190 to 1.4230 (20°C, 589 nm)
Appearance (Form)Liquid
Distillation range170 to 195 °C
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection