Ethanesulfonic acid, 95%, Thermo Scientific Chemicals
Ethanesulfonic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethanesulfonic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 366040050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS594-45-6
IUPAC Nameethanesulfonate
Molecular FormulaC2H5O3S
InChI KeyCCIVGXIOQKPBKL-UHFFFAOYSA-M
SMILESCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=94 %
Refractive index1.4325 to 1.4355 (20°C, 589 nm)
Appearance (Color)Clear yellow to brown
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only