Thermo Scientific Chemicals

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: 366390050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: 366390050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 366390050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
180516-87-4
IUPAC Name
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Molecular Formula
C13H17BO4
InChI Key
IYDKBQIEOBXLTP-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(O)=O
Appearance (Color)
White to cream
Appearance (Form)
Crystalline powder or crystals or needles
Infrared spectrum
Conforms
Melting point
227°C to 232°C
Titration with NaOH
96.0 to 104.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text