Tri-o-tolyl phosphate, 96%, Thermo Scientific Chemicals
Tri-o-tolyl phosphate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tri-o-tolyl phosphate, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 366461000
Price (USD)/ Each
134.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
158.00 
Save 23.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
134.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
158.00 
Save 23.35 (15%)
Add to cart
Tri-o-tolyl phosphate, 96%, Thermo Scientific Chemicals
Catalog number366461000
Price (USD)/ Each
134.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
158.00 
Save 23.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nametris(2-methylphenyl) phosphate
Molecular FormulaC21H21O4P
InChI KeyYSMRWXYRXBRSND-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1OP(=O)(OC1=CC=CC=C1C)OC1=CC=CC=C1C
Molecular Weight (g/mol)368.37
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Viscous liquid
Infrared spectrumConforms
GC>=95 %
Refractive index1.5580 to 1.5610 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only