Thermo Scientific Chemicals

Propargyltrimethylsilane, 80-90%, stabilized, Thermo Scientific Chemicals

Catalog number: 366630010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Propargyltrimethylsilane, 80-90%, stabilized, Thermo Scientific Chemicals

Catalog number: 366630010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 366630010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13361-64-3
IUPAC Name
trimethyl(prop-2-yn-1-yl)silane
Molecular Formula
C6H12Si
InChI Key
ULYLMHUHFUQKOE-UHFFFAOYSA-N
SMILES
C[Si](C)(C)CC#C
GC
80 to 90 %
Appearance (Form)
Liquid
Stabilizer
0.1 % hydroquinone
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text