Thermo Scientific Chemicals

Aluminum oxide, for chromatography, neutral, Brockmann I, 40-300 μm, 60A, Thermo Scientific Chemicals

Catalog number: 366680010
1 kg, Each
Thermo Scientific Chemicals

Aluminum oxide, for chromatography, neutral, Brockmann I, 40-300 μm, 60A, Thermo Scientific Chemicals

Catalog number: 366680010
1 kg, Each
Quantity
Catalog number: 366680010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1344-28-1
IUPAC Name
dialuminium(3+) trioxidandiide
Molecular Formula
Al2O3
InChI Key
PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Al+3].[Al+3]
Sodium oxide (Na2O)
=<0.4 %
Silicon dioxide (SiO2)
=<0.03 %
Appearance (Color)
White to off-white
pH
6.5 to 7.5 (10% aqueous suspension)
Particle size
=<5 % (<40 µm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for column chromatography
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text