Thermo Scientific Chemicals

1,2,3,4-Butanetetracarboxylic acid, 99+%, Thermo Scientific Chemicals

Catalog number: 366765000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

1,2,3,4-Butanetetracarboxylic acid, 99+%, Thermo Scientific Chemicals

Catalog number: 366765000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 366765000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1703-58-8
IUPAC Name
butane-1,2,3,4-tetracarboxylic acid
Molecular Formula
C8H10O8
InChI Key
GGAUUQHSCNMCAU-UHFFFAOYNA-N
SMILES
OC(=O)CC(C(CC(O)=O)C(O)=O)C(O)=O
Appearance (Color)
White to off-white
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
188°C to 192°C
Titration with NaOH
99.0 to 101.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text