Thermo Scientific Chemicals

6-Aminonicotinic acid methyl ester, 97%, Thermo Scientific Chemicals

Catalog number: 366810250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

6-Aminonicotinic acid methyl ester, 97%, Thermo Scientific Chemicals

Catalog number: 366810250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 366810250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
36052-24-1
IUPAC Name
2-amino-5-(methoxycarbonyl)pyridin-1-ium
Molecular Formula
C7H9N2O2
InChI Key
JACPDLJUQLKABC-UHFFFAOYSA-O
SMILES
COC(=O)C1=CC=C(N)[NH+]=C1
Appearance (Color)
White to yellow to light beige to pink
Infrared spectrum
Conforms
Melting point
155°C to 160°C
Appearance (Form)
Powder
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text