Cyclopropanecarboxaldehyde, 98%, Thermo Scientific Chemicals
Cyclopropanecarboxaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopropanecarboxaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 366820250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1489-69-6
IUPAC Namecyclopropanecarbaldehyde
Molecular FormulaC4H6O
InChI KeyJMYVMOUINOAAPA-UHFFFAOYSA-N
SMILESO=CC1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Infrared spectrumConforms
Refractive index1.4285 to 1.4305 (20°C, 589 nm)
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only