Thermo Scientific Chemicals

Di-tert-butyl azodicarboxylate, 97%, Thermo Scientific Chemicals

Catalog number: 366920050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Di-tert-butyl azodicarboxylate, 97%, Thermo Scientific Chemicals

Catalog number: 366920050
5 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 366920050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
870-50-8
IUPAC Name
N-{[(tert-butoxy)carbonyl]imino}(tert-butoxy)formamide
Molecular Formula
C10H18N2O4
InChI Key
QKSQWQOAUQFORH-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)N=NC(=O)OC(C)(C)C
Appearance (Color)
Yellow
Infrared spectrum
Conforms
GC
>=96.0 %
Melting point
89°C to 93°C
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X Bond Formation (Halogen), Radical initiators
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text