Thermo Scientific Chemicals

4-Hydroxycarbazole, 98+%, Thermo Scientific Chemicals

Catalog number: 366960010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Hydroxycarbazole, 98+%, Thermo Scientific Chemicals

Catalog number: 366960010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 366960010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
52602-39-8
IUPAC Name
9H-carbazol-4-ol
Molecular Formula
C12H9NO
InChI Key
UEOHATPGKDSULR-UHFFFAOYSA-N
SMILES
OC1=CC=CC2=C1C1=C(N2)C=CC=C1
HPLC
>=98.0 %
Appearance (Color)
Tan to dark brown
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
163°C to 168°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text