Ethylene diacrylate, 95%, stabilized, Thermo Scientific Chemicals
Ethylene diacrylate, 95%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethylene diacrylate, 95%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 366970050
Price (USD)/ Each
152.65
Online exclusive
179.00 
Save 26.35 (15%)
-
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
152.65
Online exclusive
179.00 
Save 26.35 (15%)
Add to cart
Ethylene diacrylate, 95%, stabilized, Thermo Scientific Chemicals
Catalog number366970050
Price (USD)/ Each
152.65
Online exclusive
179.00 
Save 26.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-(prop-2-enoyloxy)ethyl prop-2-enoate
Molecular FormulaC8H10O4
InChI KeyKUDUQBURMYMBIJ-UHFFFAOYSA-N
SMILESC=CC(=O)OCCOC(=O)C=C
Molecular Weight (g/mol)170.16
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Stabilizer100 to 400 ppm MEHQ
Appearance (Color)Clear colorless to light yellow
Refractive index1.4520 to 1.4560 (20°C, 589 nm)
GC>=94 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only