Thermo Scientific Chemicals

Methyl 2-oxocyclohexanecarboxylate, 90%, Thermo Scientific Chemicals

Catalog number: 367010100
10 mL, Glass bottle, Each
Thermo Scientific Chemicals

Methyl 2-oxocyclohexanecarboxylate, 90%, Thermo Scientific Chemicals

Catalog number: 367010100
10 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 367010100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
41302-34-5
IUPAC Name
methyl 2-oxocyclohexane-1-carboxylate
Molecular Formula
C8H12O3
InChI Key
JEENWEAPRWGXSG-UHFFFAOYNA-N
SMILES
COC(=O)C1CCCCC1=O
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=88 % may occur as tautomers
Appearance (Color)
Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text