Thermo Scientific Chemicals

2-Iodobiphenyl, 98%, Thermo Scientific Chemicals

Catalog number: 367020250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Iodobiphenyl, 98%, Thermo Scientific Chemicals

Catalog number: 367020250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 367020250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2113-51-1
IUPAC Name
2-iodo-1,1'-biphenyl
Molecular Formula
C12H9I
InChI Key
QFUYDAGNUJWBSM-UHFFFAOYSA-N
SMILES
IC1=CC=CC=C1C1=CC=CC=C1
GC
>=97.5 %
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.6600 to 1.6620 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text