Thermo Scientific Chemicals

Calcium thioglycolate trihydrate, 99%, Thermo Scientific Chemicals

Catalog number: 367031000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Calcium thioglycolate trihydrate, 99%, Thermo Scientific Chemicals

Catalog number: 367031000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 367031000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
65208-41-5
IUPAC Name
2-sulfanylacetic acid calcium trihydrate
Molecular Formula
C2H10CaO5S
InChI Key
GDECCIJZCWLFSB-UHFFFAOYSA-N
SMILES
O.O.O.[Ca].OC(=O)CS
Titration Iodometric
>=98.5 %
Appearance (Color)
White
Appearance (Form)
Powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text