p-Toluenesulfonic anhydride, 95%, Thermo Scientific Chemicals
p-Toluenesulfonic anhydride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

p-Toluenesulfonic anhydride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 367070050
Price (USD)/ Each
52.80
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
52.80
Add to cart
p-Toluenesulfonic anhydride, 95%, Thermo Scientific Chemicals
Catalog number367070050
Price (USD)/ Each
52.80
-
Add to cart
Chemical Identifiers
IUPAC Name4-methylbenzenesulfonyl 4-methylbenzene-1-sulfonate
Molecular FormulaC14H14O5S2
InChI KeyPDVFSPNIEOYOQL-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(=O)(=O)OS(=O)(=O)C1=CC=C(C)C=C1
Molecular Weight (g/mol)326.38
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
Titration by coupling>=94 %
Appearance (Color)Off-white to beige to grey
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only