Thermo Scientific Chemicals

N-(α,α,α-Trifluoro-p-tolyl)piperazine, 98%, Thermo Scientific Chemicals

Catalog number: 367120050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

N-(α,α,α-Trifluoro-p-tolyl)piperazine, 98%, Thermo Scientific Chemicals

Catalog number: 367120050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 367120050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
30459-17-7
IUPAC Name
1-[4-(trifluoromethyl)phenyl]piperazine
Molecular Formula
C11H13F3N2
InChI Key
IBQMAPSJLHRQPE-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC=C(C=C1)N1CCNCC1
Appearance (Color)
Colorless to light yellow
Appearance (Form)
Crystals
Infrared spectrum
Conforms
GC
>=97.5 %
Melting point
85°C to 90°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text