Thermo Scientific Chemicals

1-(Tetrahydro-2-furoyl)piperazine, 98%, Thermo Scientific Chemicals

Catalog number: 367160050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

1-(Tetrahydro-2-furoyl)piperazine, 98%, Thermo Scientific Chemicals

Catalog number: 367160050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 367160050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
63074-07-7
IUPAC Name
4-[(2S)-oxolane-2-carbonyl]piperazin-1-ium
Molecular Formula
C9H17N2O2
InChI Key
UKESBLFBQANJHH-QMMMGPOBSA-O
SMILES
O=C([C@@H]1CCCO1)N1CC[NH2+]CC1
GC
>=97.5 %
Appearance (Color)
Colorless to yellow
Appearance (Form)
Viscous liquid
Infrared spectrum
Conforms
Refractive index
1.5195 to 1.5215 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text