4-Amino-3,5-dichloropyridine, 97%, Thermo Scientific Chemicals
4-Amino-3,5-dichloropyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Amino-3,5-dichloropyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number 367220250
Price (USD)/ Each
99.80
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
99.80
Add to cart
4-Amino-3,5-dichloropyridine, 97%, Thermo Scientific Chemicals
Catalog number367220250
Price (USD)/ Each
99.80
-
Add to cart
Chemical Identifiers
CAS22889-78-7
IUPAC Name3,5-dichloropyridin-4-amine
Molecular FormulaC5H4Cl2N2
InChI KeyISIQAMHROGZHOV-UHFFFAOYSA-N
SMILESNC1=C(Cl)C=NC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=96.0 %
Appearance (Color)White to beige
Melting point156°C to 164°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only