3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 97%, Thermo Scientific Chemicals
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 367590010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS210907-84-9
IUPAC Name3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular FormulaC12H18BNO2
InChI KeyYMXIIVIQLHYKOT-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC=CC(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light brown
Appearance (Form)Powder or crystals
Infrared spectrumConforms
GC>=96.0 %
Melting point89°C to 94°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only