4-N-BOC-Aminopiperidine, 98%, Thermo Scientific Chemicals
4-N-BOC-Aminopiperidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-N-BOC-Aminopiperidine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Catalog number 367620050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS73874-95-0
IUPAC Name4-{[(tert-butoxy)carbonyl]amino}piperidin-1-ium
Molecular FormulaC10H21N2O2
InChI KeyCKXZPVPIDOJLLM-UHFFFAOYSA-O
SMILESCC(C)(C)OC(=O)NC1CC[NH2+]CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Melting point155°C to 160°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only