Thermo Scientific Chemicals

Methyl 2-(bromomethyl)acrylate, 96%, Thermo Scientific Chemicals

Catalog number: 367630050
5 g, Each
Thermo Scientific Chemicals

Methyl 2-(bromomethyl)acrylate, 96%, Thermo Scientific Chemicals

Catalog number: 367630050
5 g, Each
Quantity
Catalog number: 367630050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4224-69-5
IUPAC Name
methyl 2-(bromomethyl)prop-2-enoate
Molecular Formula
C5H7BrO2
InChI Key
CFTUQSLVERGMHL-UHFFFAOYSA-N
SMILES
COC(=O)C(=C)CBr
Appearance (Color)
Clear colorless to light brown
Infrared spectrum
Conforms
Appearance (Form)
Liquid
Refractive index
1.4870 to 1.4910 (20°C, 589 nm)
GC
>=95 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text