Methyl 2-(bromomethyl)acrylate, 96%, Thermo Scientific Chemicals
Methyl 2-(bromomethyl)acrylate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 2-(bromomethyl)acrylate, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number 367630250
Price (USD)/ Each
1,630.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
1,630.00
Add to cart
Methyl 2-(bromomethyl)acrylate, 96%, Thermo Scientific Chemicals
Catalog number367630250
Price (USD)/ Each
1,630.00
-
Add to cart
Chemical Identifiers
CAS4224-69-5
IUPAC Namemethyl 2-(bromomethyl)prop-2-enoate
Molecular FormulaC5H7BrO2
InChI KeyCFTUQSLVERGMHL-UHFFFAOYSA-N
SMILESCOC(=O)C(=C)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light brown
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4870 to 1.4910 (20°C, 589 nm)
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only