8-Bromooctanoic acid, 95%, Thermo Scientific Chemicals
8-Bromooctanoic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

8-Bromooctanoic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 367660100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS17696-11-6
IUPAC Name8-bromooctanoate
Molecular FormulaC8H14BrO2
InChI KeyBKJFDZSBZWHRNH-UHFFFAOYSA-M
SMILES[O-]C(=O)CCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
Appearance (Color)White to cream
Infrared spectrumConforms
Melting point36°C to 41°C
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only